missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing, Fisher Chemical™

Product Code. 10715541 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10715541 100 mL 100mL
10509210 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10715541 Supplier Fisher Chemical Supplier No. T/3255/PB05
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name 2,2,2-trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

pH 0.45
Melting Point -15°C
Boiling Point 72°C
Color Colorless
Formula Weight 114.02g/mol
Quantity 100 mL
Packaging Plastic Coated Glass bottle
Physical Form Liquid
Vapor Density 3.9
Vapor Pressure 107mbar 25
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.