missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonic Acid Magnesium Salt, TRC

Product Code. 30456539
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30456539

Brand: Toronto Research Chemicals TRCT8975735G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trifluoromethanesulfonic Acid Magnesium Salt
CAS 60871-83-2
Molecular Formula (CF3SO3)2Mg
Quantity 5 g
Synonym Magnesium bis(trifluoromethanesulfonate), Magnesium Triflate, Magnesium trifluoromethanesulfonate, Magnesium trifluoromethanesulfonate (1:2), Trifluoromethanesulfonic Acid Magnesium Salt
InChI Formula InChI=1S/2CHF3O3S.Mg/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2
SMILES [Mg+2].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
IUPAC Name magnesium;trifluoromethanesulfonate
Molecular Weight (g/mol) 322.44
Formula Weight 321.8891
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.