missing translation for 'onlineSavingsMsg'
Learn More

Triglycidyl Isocyanurate, TRC

Product Code. 30455123
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455123 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455123 Supplier Toronto Research Chemicals Supplier No. TRCT79381510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2451-62-9
Molecular Formula C12 H15 N3 O6
Quantity 10 g
Synonym 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-tris(oxiranylmethyl)- (9CI), s-Triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-tris(2,3-epoxypropyl)- (8CI), s-Triazine-2,4,6(1H,3H,5H)-trione, tris(2,3-epoxypropyl)- (7CI), 1,3,5-Tris(2-oxiranylmethyl)-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Triglycidyl isocyanurate, 1,3,5-Triglycidyl-s-triazine-2,4,6-trione, 1,3,5-Triglycidylhexahydro-1,3,5-triazine-2,4,6-trione, 1,3,5-Triglycidylisocyanuric acid, 1,3,5-Tris(2,3-epoxypropyl) isocyanurate, 1,3,5-Tris(oxiran-2-ylmethyl)-1,3,5-triazine-2,4,6-trione, 1,3,5-Tris(oxiranylmethyl)-1,3,5-triazine-2,4,6-trione, Glycidyl isocyanurate, N,N',N' '-Triglycidyl isocyanurate, NSC 269934, PTGIC, TGT, Triglycidyl isocyanurate, Tris(2,3-epoxypropyl) isocyanurate, Tris(epoxypropyl) isocyanurate
InChI Formula InChI=1S/C12H15N3O6/c16-10-13(1-7-4-19-7)11(17)15(3-9-6-21-9)12(18)14(10)2-8-5-20-8/h7-9H,1-6H2
SMILES O=C1N(CC2CO2)C(=O)N(CC3CO3)C(=O)N1CC4CO4
IUPAC Name 1,3,5-tris(oxiran-2-ylmethyl)-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol) 297.26
Formula Weight 297.0961
Chemical Name or Material 1,3,5-Triglycidyl isocyanurate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.