missing translation for 'onlineSavingsMsg'
Få mere at vide

Triglyme, TRC

Product Code. 30456396
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456396 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456396 Supplier Toronto Research Chemicals Supplier No. TRCT7938001G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Triglyme
CAS 112-49-2
Molecular Formula C8 H18 O4
Quantity 1 g
Synonym 2,5,8,11-Tetraoxadodecane, 1,2-Bis(2-methoxyethoxy)ethane, Ansul Ether 161, DMTG, Ethane','1,2-bis(2-methoxyethoxy)-, Glyme 4, Hisolve MTM, Methyltriglyme, NSC 66400, Triethylene glycol dimethyl ether, Triglyme
InChI Formula InChI=1S/C8H18O4/c1-9-3-5-11-7-8-12-6-4-10-2/h3-8H2,1-2H3
SMILES COCCOCCOCCOC
IUPAC Name 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
Molecular Weight (g/mol) 178.23
Formula Weight 178.1205
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.