missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triglyme, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Triglyme |
| CAS | 112-49-2 |
| Molecular Formula | C8 H18 O4 |
| Quantity | 25 g |
| Synonym | 2,5,8,11-Tetraoxadodecane, 1,2-Bis(2-methoxyethoxy)ethane, Ansul Ether 161, DMTG, Ethane','1,2-bis(2-methoxyethoxy)-, Glyme 4, Hisolve MTM, Methyltriglyme, NSC 66400, Triethylene glycol dimethyl ether, Triglyme |
| InChI Formula | InChI=1S/C8H18O4/c1-9-3-5-11-7-8-12-6-4-10-2/h3-8H2,1-2H3 |
| SMILES | COCCOCCOCCOC |
| IUPAC Name | 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane |
| Molecular Weight (g/mol) | 178.23 |
| Formula Weight | 178.1205 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction