missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilyl Trifluoromethanesulfonate, TRC

Product Code. 30453232
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30453232

Brand: Toronto Research Chemicals TRCT7905505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triisopropylsilyl Trifluoromethanesulfonate
CAS 80522-42-5
Molecular Formula C10H21F3O3SSi
Quantity 5 g
Synonym TIPSOTf, Triisopropyl(trifluoromethanesulfonato)silane, Triisopropylsilyl Triflate, TIPS Triflate, 1,1,1-Trifluoromethanesulfonic Acid Tris(1-methylethyl)silyl Ester
InChI Formula InChI=1S/C10H21F3O3SSi/c1-7(2)18(8(3)4,9(5)6)16-17(14,15)10(11,12)13/h7-9H,1-6H3
SMILES CC(C)[Si](OS(=O)(=O)C(F)(F)F)(C(C)C)C(C)C
IUPAC Name tri(propan-2-yl)silyl trifluoromethanesulfonate
Molecular Weight (g/mol) 306.42
Formula Weight 306.0933
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.