Learn More
TRIM21 Mouse anti-Human, Unconjugated, Clone: 1E8B9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67136-1-IG-150UL
 View more versions of this product
Product Code. 16819595
  • 410.00€ / 150µL
Description
The TRIM21 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM21. This antibody recognizes human, mouse, rat antigen. The TRIM21 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TRIM21
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunoprecipitation, Immunohistochemistry
1E8B9
Unconjugated
150 μL
6737