missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM31 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93541-0.1ml
This item is not returnable.
View return policy
Description
TRIM31 Polyclonal antibody specifically detects TRIM31 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| TRIM31 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| C6orf13E3 ubiquitin-protein ligase TRIM31, EC 6.3.2.-, HCG1, HCGItripartite motif-containing 31 gamma, RNFring finger protein, tripartite motif containing 31, tripartite motif-containing 31, Tripartite motif-containing protein 31 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 186-425 of human TRIM31 (NP_008959.3). ELLHQVLEEEKNFLLSRIYWLGHEGTEAGKHYVASTEPQLNDLKKLVDSLKTKQNMPPRQLLEDIKVVLCRSEEFQFLNPTPVPLELEKKLSEAKSRHDSITGSLKKFKDQLQADRKKDENRFFKSMNKNDMKSWGLLQKNNHKMNKTSEPGSSSAGGRTTSGPPNHHSSAPSHSLFRASSAGKVTFPVCLLASYDEISGQGASSQDTKTFDVALSEELHAALSEWLTAIRAWFCEVPSS | |
| 0.1 mL | |
| Zinc Finger | |
| 11074 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction