Learn More
TRIM32 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-10326-20UL
Description
The TRIM32 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRIM32. This antibody recognizes human, mouse, rat antigen. The TRIM32 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| TRIM32 | |
| 1000 μg/mL | |
| 72 kDa Tat interacting protein, BBS11, HT2A, LGMD2H, TATIP, TRIM32, tripartite motif containing 32, Zinc finger protein HT2A | |
| 20 μL | |
| 22954 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |