Learn More
TRIM32 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-10326-20UL
 View more versions of this product
Product Code. 18045255
  • 145.00€ / 20µL
Description
The TRIM32 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRIM32. This antibody recognizes human, mouse, rat antigen. The TRIM32 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
TRIM32
1000 μg/mL
72 kDa Tat interacting protein, BBS11, HT2A, LGMD2H, TATIP, TRIM32, tripartite motif containing 32, Zinc finger protein HT2A
20 μL
22954
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa