Learn More
TRIM54 Monoclonal Antibody (OTI6C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803873
Description
The protein encoded by this gene contains a RING finger motif and is highly similar to the ring finger proteins RNF28/MURF1 and RNF29/MURF2. In vitro studies demonstrated that this protein, RNF28, and RNF29 form heterodimers, which may be important for the regulation of titin kinase and microtubule-dependent signal pathways in striated muscles. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
| TRIM54 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BYV2 | |
| TRIM54 | |
| Human recombit protein fragment corresponding to amino acids 166-400 of human TRIM54 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6C3 | |
| Unconjugated | |
| TRIM54 | |
| MURF, MURF-3, RNF30, muRF3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57159 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.