Learn More
TRIM72 Mouse, anti-Human, CoraLite Plus 488, Clone: 5F2C11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67403-20UL
 View more versions of this product
Product Code. 18087415
  • 145.00€ / 20µL
Description
The TRIM72 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM72. This antibody recognizes human, pig, mouse antigen. The TRIM72 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
TRIM72
5F2C11
CoraLite Plus 488
Mouse
Primary
Human, Pig, Mouse
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
hMG53, Mg53, Mitsugumin 53, TRIM72, tripartite motif containing 72
20 μL
493829
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.