missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimebutine Maleate, LoGiCal
Description
Trimebutine Maleate
Specifications
Specifications
| Chemical Name or Material | Trimebutine Maleate |
| CAS | 34140-59-5 |
| Analyte or Component Names | Trimebutine Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H29 N O5 . C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Trimebutine Maleate, Benzoic acid, 3,4,5-trimethoxy-, 2-(dimethylamino)-2-phenylbutyl ester, (2 Z)-2-butenedioate (1:1) (9 CI), Benzoic acid, 3,4,5-trimethoxy-, 2-(dimethylamino)-2-phenylbutyl ester, (Z)-2-butenedioate (1:1), Cerekinon, Debridat, Benzoic acid, 3,4,5-trimethoxy-, beta-(dimethylamino)-beta-ethylphenethyl ester maleate (1:1) (8 CI), Benzoic acid, 3,4,5-trimethoxy-, beta-(dimethylamino)-beta-ethylphenethyl ester maleate (1:1) (8 CI), Benzoic acid, 3,4,5-trimethoxy-, beta-(dimethylamino)-beta-ethylphenethyl ester maleate (1:1) (8 CI), Benzoic acid, 3,4,5-trimethoxy-, beta-(dimethylamino)-beta-ethylphenethyl ester maleate (1:1) (8 CI), Benzoic acid, 3,4,5-trimethoxy-, beta-(dimethylamino)-beta-ethylphenethyl ester maleate (1:1) (8 CI), Benzoic acid, 3,4,5-trimethoxy-, beta-(dimethylamino)-beta-ethylphenethyl ester maleate (1:1) (8 CI), Benzoic acid, 3,4,5-trimethoxy-, beta-(dimethylamino)-beta-ethylphenethyl ester maleate (1:1) (8 CI), Benzoic acid, 3,4,5-trimethoxy-, beta-(dimethylamino)-beta-ethylphenethyl ester maleate (1:1) (8 CI), Benzoic acid, 3,4,5-trimethoxy-, beta-(dimethylamino)-beta-ethylphenethyl ester maleate (1:1) (8 CI) |
| InChI Formula | InChI=1 S/C22H29NO5.C4H4O4/c1-7-22(23(2)3,17-11-9-8-10-12-17)15-28-21(24)16-13-18(25-4)20(27-6)19(14-16)26-5;5-3(6)1-2-4(7)8/h8-14 H,7,15H2,1-6H3;1-2 H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(N(C)C)c2ccccc2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;[2-(dimethylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?