Learn More
Trimebutine Maleate Salt, TRC
Product Code. 17937814
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17937814 5 g 5g
17968494 2.5 g 2.5g
17978674 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17937814 Supplier Toronto Research Chemicals Supplier No. TRCT7956055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34140-59-5
Molecular Formula C22 H29 N O5 . C4 H4 O4
Molecular Weight (g/mol) 503.54
Synonym Trimebutine Maleate, Benzoic acid, 3,4,5-trimethoxy-, 2-(dimethylamino)-2-phenylbutyl ester, (2Z)-2-butenedioate (1:1) (9CI), Benzoic acid, 3,4,5-trimethoxy-, β-(dimethylamino)-β-ethylphenethyl ester maleate (1:1) (8CI), Benzoic acid, 3,4,5-trimethoxy-, 2-(dimethylamino)-2-phenylbutyl ester, (Z)-2-butenedioate (1:1), Cerekinon, Debridat, Digerent, Dromostat, Foldox, Polibutin, Spabucol, TM 906, Trimedat
IUPAC Name (Z)-but-2-enedioic acid;[2-(dimethylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate
SMILES CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(N(C)C)c2ccccc2.OC(=O)\C=C/C(=O)O
Especificaciones
Quantity 5 g
InChI Formula InChI=1S/C22H29NO5.C4H4O4/c1-7-22(23(2)3,17-11-9-8-10-12-17)15-28-21(24)16-13-18(25-4)20(27-6)19(14-16)26-5;5-3(6)1-2-4(7)8/h8-14H,7,15H2,1-6H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formula Weight 503.2155
Percent Purity >95%
Chemical Name or Material Trimebutine Maleate
Grade HPLC
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.