missing translation for 'onlineSavingsMsg'
Learn More

Trimebutine Maleate Salt, TRC

Product Code. 17978674
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17978674 1 g 1g
17937814 5 g 5g
17968494 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17978674 Supplier Toronto Research Chemicals Supplier No. TRCT7956051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34140-59-5
Molecular Formula C22 H29 N O5 . C4 H4 O4
Molecular Weight (g/mol) 503.54
Synonym Trimebutine Maleate, Benzoic acid, 3,4,5-trimethoxy-, 2-(dimethylamino)-2-phenylbutyl ester, (2Z)-2-butenedioate (1:1) (9CI), Benzoic acid, 3,4,5-trimethoxy-, β-(dimethylamino)-β-ethylphenethyl ester maleate (1:1) (8CI), Benzoic acid, 3,4,5-trimethoxy-, 2-(dimethylamino)-2-phenylbutyl ester, (Z)-2-butenedioate (1:1), Cerekinon, Debridat, Digerent, Dromostat, Foldox, Polibutin, Spabucol, TM 906, Trimedat
IUPAC Name (Z)-but-2-enedioic acid;[2-(dimethylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate
SMILES CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(N(C)C)c2ccccc2.OC(=O)\C=C/C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H29NO5.C4H4O4/c1-7-22(23(2)3,17-11-9-8-10-12-17)15-28-21(24)16-13-18(25-4)20(27-6)19(14-16)26-5;5-3(6)1-2-4(7)8/h8-14H,7,15H2,1-6H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formula Weight 503.2155
Percent Purity >95%
Chemical Name or Material Trimebutine Maleate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.