missing translation for 'onlineSavingsMsg'
Learn More

Trimethadione, TRC

Product Code. 30456897
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
1g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456897 2.5 g 2.5g
30449895 25 g 25g
30450132 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456897 Supplier Toronto Research Chemicals Supplier No. TRCT7961552.5G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-48-0
Molecular Formula C6 H9 N O3
Molecular Weight (g/mol) 143.14
Synonym Trimethadione, 3,5,5-Trimethyloxazolidine-2,4-dione, 3,3,5-Trimethyl-2,4-diketooxazolidine, 3,5,5-Trimethyl-2,4-oxazolidinedione, Absentol, Edion, Epidione, Epixal, Minoaleviatin, NSC 15799, NSC 169503, Petidion, Petidon, Petilep, Pitmal, Ptimal, Tridion, Tridione, Tridone, Trimedal, Trimetadione, Trimethadion, Trimethadione, Trimethin, Trimetin, Trioxanona, Troxidone, 2,4-Oxazolidinedione, 3,5,5-trimethyl-
IUPAC Name 3,5,5-trimethyl-1,3-oxazolidine-2,4-dione
SMILES CN1C(=O)OC(C)(C)C1=O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C6H9NO3/c1-6(2)4(8)7(3)5(9)10-6/h1-3H3
Formula Weight 143.0582
Chemical Name or Material Trimethadione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.