missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(2-phenylethyl)silane, TRC

Product Code. 30457318
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457318 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457318 Supplier Toronto Research Chemicals Supplier No. TRCT7974552.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethoxy(2-phenylethyl)silane
CAS 49539-88-0
Molecular Formula C11H18O3Si
Quantity 2.5 g
Synonym [2-(Trimethoxysilyl)ethyl]benzene, (2-Phenylethyl)trimethoxysilane, P 113, P 113 (Silane Derivative), Phenethyltrimethoxysilane, SIP 6722.6, T 2869, β-(Trimethoxysilyl)ethylbenzene
InChI Formula InChI=1S/C11H18O3Si/c1-12-15(13-2,14-3)10-9-11-7-5-4-6-8-11/h4-8H,9-10H2,1-3H3
SMILES CO[Si](CCC1=CC=CC=C1)(OC)OC
IUPAC Name trimethoxy(phenethyl)silane
Molecular Weight (g/mol) 226.34
Formula Weight 226.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.