missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(octyl)silane, TRC

Product Code. 30452013
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452013 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452013 Supplier Toronto Research Chemicals Supplier No. TRCT773450250G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethoxy(octyl)silane
CAS 3069-40-7
Molecular Formula C11 H26 O3 Si
Quantity 250 g
Synonym Trimethoxyoctylsilane, Dynasylan OCTMO, JH-N 318, KBM 3083, Octyltrimethoxysilane, Si 108, Sil-Act ATS 100LV, Sil-Act ATS 22, Sil-Act ATS 42A, Sil-Act ATS 42NS, Silan 108, XC 95-738, n-Octyltrimethoxysilane
InChI Formula InChI=1S/C11H26O3Si/c1-5-6-7-8-9-10-11-15(12-2,13-3)14-4/h5-11H2,1-4H3
SMILES CCCCCCCC[Si](OC)(OC)OC
IUPAC Name trimethoxy(octyl)silane
Molecular Weight (g/mol) 234.408
Formula Weight 234.165
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.