missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxyvinylsilane, Dr. Ehrenstorfer

Product Code. 30309060
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309060 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309060 Supplier Dr. Ehrenstorfer Supplier No. DRECA17876900
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethoxyvinylsilane
CAS 2-7-2768
Analyte or Component Names Trimethoxyvinylsilane
Shipping Condition +20°C
Molecular Formula C5 H12 O3 Si
Certifications/Compliance ISO 17034
Synonym Silane, ethenyltrimethoxy-, Silane, trimethoxyvinyl- (6CI,7CI,8CI), Ethenyltrimethoxysilane, (Trimethoxysilyl)ethene, A 101, A 101 (silane), A 171, AC 71, CV 4917, DB 171, DL 171, Dow Corning 6300, Dynasylan Si 108, Dynasylan Silfin, Dynasylan Silfin 24, Dynasylan VTMO, EMI 1833, GX 171, Geniosil X 10GS, Geniosil XL 10, KBM 1003, KBM 3100, KH 171, KH 921, KHA 171, LMD 171, LS 815, LT 171, NUC Silicone A 171, NUC-Y 9818, Penta 1002, Q 9-6300, S 210, SB 6301, SCA 1603, SG-Si 171, SH 6300, SIV 9220.0, SL 815, SZ 6300, Sigma T 5051, Sila-Ace 210, Sila-Ace S 210, Silane A 171, Silane V Trimethoxy, Silaplane S 210, Silfin 13, Silfin 22, Silfin 24, Silfin 59, Silopren TP 3625, Silox VS 911, Silquest A 171, TSL 8310, U 611, UN 1993, UP 171, V 0042, V 06, V 06 (coupling agent), V 4917, VTMO, VTMS, VTS-M, Vinyltrimethoxysilane, WD 21, XL 10, XL 10 (silane), Xiameter OFS 6300, Y 4302, YDH 171, YGO 1202, Z 6300
InChI Formula InChI=1S/C5H12O3Si/c1-5-9(6-2,7-3)8-4/h5H,1H2,2-4H3
SMILES CO[Si](OC)(OC)C=C
IUPAC Name ethenyl(trimethoxy)silane
Molecular Weight (g/mol) 148.2325
Quantity 1 mL
Formula Weight 148.0556 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.