missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl[3-(perfluorooctanamido)propyl]ammonium Iodide, TRC

Product Code. 30417257
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417257 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417257 Supplier Toronto Research Chemicals Supplier No. TRCP229740100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[(Perfluorooxooctyl)amino]propyltrimethylammoniumiodide
CAS 335-90-0
Molecular Formula C14 H16 F15 N2 O . I
Quantity 100 mg
Synonym Trimethyl[3-(2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanamido)propyl]ammonium Iodide
InChI Formula InChI=1S/C14H15F15N2O.HI/c1-31(2,3)6-4-5-30-7(32)8(15,16)9(17,18)10(19,20)11(21,22)12(23,24)13(25,26)14(27,28)29;/h4-6H2,1-3H3;1H
SMILES O=C(C(F)(C(F)(C(F)(C(F)(C(F)(C(F)(C(F)(F)F)F)F)F)F)F)F)NCCC[N+](C)(C)C.[I-]
IUPAC Name N,N,N-trimethyl-3-(2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanamido)propan-1-aminium iodide
Molecular Weight (g/mol) 640.17
Formula Weight 640.007
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.