missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthoacetate, TRC

Product Code. 30457000
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457000 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457000 Supplier Toronto Research Chemicals Supplier No. TRCT7965055ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethyl Orthoacetate
CAS 1445-45-0
Purity Grade Notes GC
Molecular Formula C5H12O3
Quantity 5 mL
Synonym Orthoacetic Acid Trimethyl Ester, 1,1,1-Trimethoxyethane, Methyl Orthoacetate
InChI Formula InChI=1S/C5H12O3/c1-5(6-2,7-3)8-4/h1-4H3
SMILES COC(C)(OC)OC
IUPAC Name 1,1,1-trimethoxyethane
Molecular Weight (g/mol) 120.15
Formula Weight 120.0786
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.