missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthomethoxyacetate, TRC

Product Code. 30457258
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
missing translation for 'unitSize'
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457258 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30457258 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT7981501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethyl Orthomethoxyacetate
CAS 34359-77-8
Molecular Formula C6H14O4
Quantity 1 g
Synonym Methoxyorthoacetic Acid Trimethyl Ester, 1,1,1,2-Tetramethoxyethane
InChI Formula InChI=1S/C6H14O4/c1-7-5-6(8-2,9-3)10-4/h5H2,1-4H3
SMILES COCC(OC)(OC)OC
IUPAC Name 1,1,1,2-tetramethoxyethane
Molecular Weight (g/mol) 150.17
Formula Weight 150.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.