missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetic Anhydride, TRC

Product Code. 30451866
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451866 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451866 Supplier Toronto Research Chemicals Supplier No. TRCT79576025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylacetic Anhydride
CAS 1538-75-6
Molecular Formula C10H18O3
Quantity 25 g
Synonym 2,2-Dimethylpropanoic Acid 1,1'-Anhydride, Pivalic Anhydride, 2,2,2-Trimethylacetic Anhydride, 2,2-Dimethylpropanoyl Anhydride, Neopentanoic Anhydride, Pivalic Acid Anhydride, Pivaloyl Anhydride
InChI Formula InChI=1S/C10H18O3/c1-9(2,3)7(11)13-8(12)10(4,5)6/h1-6H3
SMILES CC(C)(C)C(=O)OC(=O)C(C)(C)C
IUPAC Name 2,2-dimethylpropanoyl 2,2-dimethylpropanoate
Molecular Weight (g/mol) 186.25
Formula Weight 186.1256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.