Learn More
Alfa Aesar™ Trimethylphosphine sulfide, 97%
Product Code. 11422778
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422778 1 g 1g
11432778 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11422778 Supplier Alfa Aesar™ Supplier No. A16762.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Kemiska identifierare
CAS 2404-55-9
Molecular Formula C3H9PS
Molecular Weight (g/mol) 108.14
MDL Number MFCD00014460
InChI Key KTFAZNVGJUIWJM-UHFFFAOYSA-N
Synonym trimethylphosphine sulfide, phosphine sulfide, trimethyl, trimethyl-??-phosphanethione, acmc-20ajda, me3ps, trimethylphosphinesulfide, trimethyl sulfanylidene, phosphorane, trimethyl-, sulfide, inchi=1/c3h9ps/c1-4 2,3 5/h1-3h, dimethyl sulfanylidene-??-phosphanyl methyl
PubChem CID 123160
IUPAC Name trimethyl(sulfanylidene)-$l^{5}-phosphane
SMILES CP(C)(C)=S
Specifikationer
Melting Point 155°C to 156°C
Quantity 1 g
UN Number UN3464
Formula Weight 108.15
Percent Purity 97%
Chemical Name or Material Trimethylphosphine sulfide
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.