missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trimethylphosphine sulfide, 97%

Product Code. 11432778
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432778 5 g 5g
11422778 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432778 Supplier Alfa Aesar™ Supplier No. A16762.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2404-55-9
Molecular Formula C3H9PS
Molecular Weight (g/mol) 108.14
MDL Number MFCD00014460
InChI Key KTFAZNVGJUIWJM-UHFFFAOYSA-N
Synonym trimethylphosphine sulfide, phosphine sulfide, trimethyl, trimethyl-??-phosphanethione, acmc-20ajda, me3ps, trimethylphosphinesulfide, trimethyl sulfanylidene, phosphorane, trimethyl-, sulfide, inchi=1/c3h9ps/c1-4 2,3 5/h1-3h, dimethyl sulfanylidene-??-phosphanyl methyl
PubChem CID 123160
IUPAC Name trimethyl(sulfanylidene)-$l^{5}-phosphane
SMILES CP(C)(C)=S

Specifications

Melting Point 155°C to 156°C
Quantity 5 g
UN Number UN3464
Formula Weight 108.15
Percent Purity 97%
Chemical Name or Material Trimethylphosphine sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.