missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl bromoacetate, 98%, ACROS Organics™

Product Code. 10582231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10582231 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10582231 Supplier Thermo Scientific Acros Supplier No. 215990050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18291-80-0
Color Yellow
Density 1.28g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H11BrO2Si
Linear Formula BrCH2CO2Si(CH3)3
Refractive Index 1.4420 to 1.4450 (20°C, 589nm)
MDL Number MFCD00000187
Quantity 5 g
Synonym trimethylsilyl bromoacetate, bromoacetic acid trimethylsilyl ester, acetic acid, bromo-, trimethylsilyl ester, acmc-209ek0, trimethylsilyl monobromoacetate, trimethylsilylbromoacetate, bromoacetic acid, tms derivative, zchhfmwudhxpfn-uhfffaoysa
InChI Key ZCHHFMWUDHXPFN-UHFFFAOYSA-N
SMILES C[Si](C)(C)OC(=O)CBr
IUPAC Name trimethylsilyl 2-bromoacetate
Molecular Weight (g/mol) 211.13
PubChem CID 87554
Formula Weight 211.14
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Trimethylsilyl bromoacetate, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.