missing translation for 'onlineSavingsMsg'
Learn More

Trinexapac-ethyl, TRC

Product Code. 17922185
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922185 250 mg 250mg
17953795 1 g 1g
17970945 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17922185 Supplier Toronto Research Chemicals Supplier No. TRCT886298250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95266-40-3
Molecular Formula C13 H16 O5
Molecular Weight (g/mol) 252.26
Synonym Cyclohexanecarboxylic acid, 4-(cyclopropylhydroxymethylene)-3,5-dioxo-, ethyl ester (9CI, ACI), CGA 163935, Cimectacarb, Moddus, Primo, Primo Maxx, Trinexapac-ethyl
IUPAC Name ethyl 4-[cyclopropyl(hydroxy)methylidene]-3,5-dioxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CC(=O)C(=C(O)C2CC2)C(=O)C1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H16O5/c1-2-18-13(17)8-5-9(14)11(10(15)6-8)12(16)7-3-4-7/h7-8,16H,2-6H2,1H3/b12-11-
Formula Weight 252.0998
Chemical Name or Material Trinexapac-ethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.