missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine Hydrobromide, TRC

Product Code. 30455940
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455940 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455940 Supplier Toronto Research Chemicals Supplier No. TRCT88650050G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triphenylphosphine Hydrobromide
CAS 6399-81-1
Molecular Formula C18H16BrP
Quantity 50 g
Synonym Triphenylphosphine compd. with Hydrobromic Acid, Triphenylphosphonium bromide
InChI Formula InChI=1S/C18H15P.BrH/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h1-15H;1H
SMILES Br.c1ccc(cc1)P(c2ccccc2)c3ccccc3
IUPAC Name triphenylphosphane;hydrobromide
Molecular Weight (g/mol) 343.2
Formula Weight 342.0173
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.