missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine sulfide, 98%, Thermo Scientific Chemicals

Product Code. 11422918
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422918 25 g 25g
11432918 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422918 Supplier Thermo Scientific Alfa Aesar Supplier No. A16884.14
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Triphenylphosphine sulfide involves in the , conversion of epoxides to episulfides (thiiranes) in presence of TFA of the same stereochemistry. It acts as a ligand for the Pd-catalyzed bisalkoxycarbonylation of olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylphosphine sulfide involves in the , conversion of epoxides to episulfides (thiiranes) in presence of TFA of the same stereochemistry. It acts as a ligand for the Pd-catalyzed bisalkoxycarbonylation of olefins.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3878-45-3
Molecular Formula C18H15PS
Molecular Weight (g/mol) 294.35
MDL Number MFCD00004917
InChI Key VYNGFCUGSYEOOZ-UHFFFAOYSA-N
Synonym triphenylphosphine sulfide, phosphine sulfide, triphenyl, triphenylphosphine sulphide, triphenylphosphine monosulfide, triphenylphosphane sulfide, triphenylphosphino-1-thione, triphenylthioxophosphorane, triphenylphosphorus sulfide, wln: spr&r&r, triphenyl-, e5-phosphanethione
PubChem CID 19758
IUPAC Name triphenyl(sulfanylidene)-$l^{5}-phosphane
SMILES S=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 158°C to 164°C
Quantity 25 g
UN Number UN3464
Beilstein 645872
Formula Weight 294.36
Percent Purity 98%
Chemical Name or Material Triphenylphosphine sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.