missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltetrazolium Chloride, Serva Electrophoresis™

Product Code. 11797846
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11797846 10 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11797846 Supplier Serva Electrophoresis Supplier No. 3713003
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 298-96-4
Melting Point >235°C
Color Cream-White
Molecular Formula C19H15ClN4
MDL Number MFCD00011963
Quantity 10 g
Synonym 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, tetrazolium chloride, 2,3,5-triphenyl-2h-tetrazol-3-ium chloride
Solubility Information 30g/L
InChI Key PKDBCJSWQUOKDO-UHFFFAOYSA-M
SMILES [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol) 334.81
PubChem CID 9283
ChEBI CHEBI:78019
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Triphenyltetrazolium chloride
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.