missing translation for 'onlineSavingsMsg'
Learn More

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III), TRC

Product Code. 30457334
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457334 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457334 Supplier Toronto Research Chemicals Supplier No. TRCT7945581G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)
CAS 17631-68-4
Molecular Formula C30 H30 Eu F21 O6
Quantity 1 g
Synonym Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')-Europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-κO,κO')-Europium, 3,5-Octanedione','6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-','europium complex Europium tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octadionate), Europium(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedione), Tris(6,6,7,7,8,8,8-hetafluoro-2,2-dimethyl-3,5-octanedionato)europium','Tris(7,7-dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedionato)europium, NSC 379435, Sievers' reagent, Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionato)Eu(III), Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionato)europium, Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedione)europium, Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedione)europium(III), Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyloctane-4,6-dionato)europium(III), Tris(2,2-dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedionato)europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III), Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-κO,κO')europium, Tris(7,7-dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedionato)europium, Tris(perfluorobutanoylpivaloylmethano)europium
InChI Formula InChI=1S/3C10H10F7O2.Eu/c3*1-7(2,3)5(18)4-6(19)8(11,12)9(13,14)10(15,16)17;/h3*4H,1-3H3;/q3*-1;+3
SMILES CC(C)(C)C1=[O][Eu+3]23([O]=C([CH-]C(=[O]2)C(F)(F)C(F)(F)C(F)(F)F)C(C)(C)C)([O]=C([CH-]C(=[O]3)C(F)(F)C(F)(F)C(F)(F)F)C(C)(C)C)[O]=C([CH-]1)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 1037.486
Formula Weight 1038.092
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.