missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium(0)-chloroform Adduct, TRC

Product Code. 30456255
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456255 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456255 Supplier Toronto Research Chemicals Supplier No. TRCT87635010G
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris(dibenzylideneacetone)dipalladium(0)-chloroform Adduct
CAS 52522-40-4
Molecular Formula C52 H43 Cl3 O3 Pd2
Quantity 10 g
Synonym Tris[μ-[(1,2-η:4,5-η)-(1E,4E)-1,5-diphenyl-1,4-pentadien-3-one]]di-palladium Compound With Trichloromethane, (E,E)-1,5-Diphenyl-1,4-pentadien-3-one Palladium Complex, Tris(dibenzylideneacetone)dipalladium Chloroform, Tris(dibenzylideneacetone)dipalladium Chloroform Adduct, Tris(dibenzylideneacetone)dipalladium(0) Chloroform Adduct, Tris(dibenzylideneacetone)dipalladium-chloroform (1:1), Tris[μ-[(1,2-η:4,5-η)-(1E,4E)-1,5-diphenyl-1,4-pentadien-3-one]]dipalladium-trichloromethane
InChI Formula InChI=1S/3C17H14O.CHCl3.2Pd/c3*18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16;2-1(3)4;;/h3*1-14H;1H;;/b3*13-11+,14-12+;;;
SMILES [Pd].[Pd].ClC(Cl)Cl.O=C(/C=C/c1ccccc1)\C=C\c2ccccc2.O=C(/C=C/c3ccccc3)\C=C\c4ccccc4.O=C(\C=C\c5ccccc5)/C=C/c6ccccc6
Molecular Weight (g/mol) 1035.1
Formula Weight 1032.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.