missing translation for 'onlineSavingsMsg'
Learn More

Tris(ethylenediamine)rhodium(III) chloride trihydrate, Rh 23.2%

Product Code. p-7048443
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11316068 0.5 g 0.5g
11336068 1 g 1g
11326068 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11316068 Supplier Thermo Scientific Alfa Aesar Supplier No. 010553.02
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15004-86-1 | C6H30Cl3N6O3Rh | 443.60 g/mol

Used as precursor for supported RH catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15004-86-1
Molecular Formula C6H30Cl3N6O3Rh
Molecular Weight (g/mol) 443.60
MDL Number MFCD00149661
InChI Key HHSLUQMEKOPKJG-UHFFFAOYSA-K
Synonym Trichlorotris(ethylenediamine)rhodium(III)
IUPAC Name rhodium(3+) tris(ethane-1,2-diamine) trihydrate trichloride
SMILES O.O.O.[Cl-].[Cl-].[Cl-].[Rh+3].NCCN.NCCN.NCCN

Specifications

Melting Point 280°C (decomposition)
Color White
Linear Formula Rh(H2NCH2CH2NH2)3Cl3·3H2O
Quantity 0.5 g
Formula Weight 443.61 (389.56 Anhydrous)
Concentration or Composition (by Analyte or Components) Rh 23.2%
Physical Form Crystalline/Fused Lump, Crystals Fuse Over Time
Chemical Name or Material Tris(ethylenediamine)rhodium(III) chloride trihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.