missing translation for 'onlineSavingsMsg'
Learn More

Tris(triphenylphosphine)rhodium(I) Chloride, TRC

Product Code. 30449707
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449707 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449707 Supplier Toronto Research Chemicals Supplier No. TRCT886465250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris(triphenylphosphine)rhodium(I) Chloride
CAS 14694-95-2
Molecular Formula C54H45ClP3Rh
Quantity 250 mg
Synonym Chlorotris(triphenylphosphine)rhodium, Chlorotris(triphenylphosphine)rhodium(I), NSC 124140, Rhodium tris(triphenylphosphine) chloride, Tris(triphenylphosphine)chlororhodium, Tris(triphenylphosphine)chlororhodium(I), Tris(triphenylphosphine)rhodium(I) Chloride, Wilkinson's Catalyst, Wilkinson's Complex
InChI Formula InChI=1S/3C18H15P.ClH.Rh/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h3*1-15H;1H;/q;;;;+1/p-1
SMILES Cl[Rh].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9
IUPAC Name chlororhodium;triphenylphosphane
Molecular Weight (g/mol) 925.21
Formula Weight 924.1478
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.