missing translation for 'onlineSavingsMsg'
Learn More

Tristearin, TRC

Product Code. 30454828
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454828 50 mg 50mg
30457020 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454828 Supplier Toronto Research Chemicals Supplier No. TRCT88645050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-43-1
Molecular Formula C57 H110 O6
Molecular Weight (g/mol) 891.48
Synonym Octadecanoic acid, 1,2,3-propanetriyl ester (9CI), Stearin, tri- (8CI), A 3D, Coatex 21, Daiwax STG, Dynasan 118, Edenor NHTi, Edenor NHTl, Esteride G-S, Glycerin tristearate, Glycerine tristearate, Glycerol trioctadecanoate, Glycerol tristearate, Glyceryl trioctadecanoate, Glyceryl tristearate, Glycolube TS, Glycowax S 932, Hardened Oil, Ligalub GT, Loxiol EP 218, PPM 17000, Pationic 919, Prisorine 2041, Radia 7512, Rikemal VT, Rikemal VT 50, SSS, Spezialfett 118, Stearic acid triglyceride, Stearic acid triglycerin ester, Stearic triglyceride, Stearoyl triglyceride, Tegin BL 150V, Triglyceride SSS, Triglyceride StStSt, Trioctadecanoin, Tristearin, Tristearoylglycerol
IUPAC Name 2,3-di(octadecanoyloxy)propyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C57H110O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h54H,4-53H2,1-3H3
Formula Weight 890.8302
Chemical Name or Material Tristearin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.