missing translation for 'onlineSavingsMsg'
Learn More

Triticonazole, TRC

Product Code. 30133143
Change view
Click to view available options
Quantity:
5 g
10 g
missing translation for 'unitSize'
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133143 10 g 10g
17974355 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30133143 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT89150310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131983-72-7
Molecular Formula C17 H20 Cl N3 O
Molecular Weight (g/mol) 317.81
Synonym (5E)-5-[(4-Chlorophenyl)methylene]-2,2-dimethyl-1-(1H-1,2,4-triazol-1-ylmethyl)cyclopentanol (ACI), (E)-(4-Chlorobenzylidene)-2,2-dimethyl-1-[1,2,4]triazol-1-ylmethyl-cyclopentanol, Alios, Charter, Premis, Premis 25 FS, Premis T, Premise, Premise (fungicide), Trinity, Triticonazole, Cyclopentanol, 5-[(4-chlorophenyl)methylene]-2,2-dimethyl-1-(1H-1,2,4-triazol-1-ylmethyl)-, (E)-, Cyclopentanol, 5-[(4-chlorophenyl)methylene]-2,2-dimethyl-1-(1H-1,2,4-triazol-1-ylmethyl)-, (E)-(±)- (ZCI), Cyclopentanol, 5-[(4-chlorophenyl)methylene]-2,2-dimethyl-1-(1H-1,2,4-triazol-1-ylmethyl)-, (5E)- (9CI, ACI)
IUPAC Name (5E)-5-[(4-chlorophenyl)methylidene]-2,2-dimethyl-1-(1,2,4-triazol-1-ylmethyl)cyclopentan-1-ol
SMILES CC1(C)CC\C(=C/c2ccc(Cl)cc2)\C1(O)Cn3cncn3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C17H20ClN3O/c1-16(2)8-7-14(9-13-3-5-15(18)6-4-13)17(16,22)10-21-12-19-11-20-21/h3-6,9,11-12,22H,7-8,10H2,1-2H3/b14-9+
Formula Weight 317.1295
Chemical Name or Material Triticonazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.