missing translation for 'onlineSavingsMsg'
Learn More

Melford Triton X-114 5 pack

Product Code. 30018907
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018907 1 Ea. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018907 Supplier Melford Supplier No. T600871

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Peroxide free detergent in single use ampoules.

Peroxide free detergent in single use ampoules. Ideal for protein isolation and sample preparation. Peroxide content: >1μg/mL. Supplied 5 ampoules per pack. Protecomics Grade

Specifications

Chemical Name or Material Triton X-114
CAS 9036-19-5, 7732-18-5
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.