missing translation for 'onlineSavingsMsg'
Learn More

TrkC Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A7F11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66380-1-IG-150UL

 View more versions of this product

Product Code. 16848205

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRKC, a member of the insulin receptor subfamily of tyrosine protein kinases, is a receptor for neurotrophin-3 (NT-3). Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.
Specifications

Specifications

TrkC
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P24786, Q03351, Q16288, Q6VNS1
Ntrk3
TrkC Fusion Protein Ag2635
150 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG1
Western Blot
2A7F11
Unconjugated
NTRK3
GP145 TrkC, gp145(trkC), NT 3 growth factor receptor, NTRK3, Trk C, TRKC, TrkC tyrosine kinase
Mouse
Protein G
RUO
18213, 29613, 397511, 4916
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.