missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Troglitazone
Description
Selective PPARγ receptor agonist (EC50 values are 780 and 555 nM at murine and human PPARγ receptors respectively). Displays no activity at PPARα or PPARδ receptors. Antidiabetic agent; exerts potent glucose-lowering effects in insulin-resistant diabetic mice. Displays anti-invasive effect on human breast cancer cells; reduces migration, adhesion and spreading on fibronectin-coated plates. Also inhibits cell growth of hematopoietic cell lines. Inhibits lamellipodia formation and actin polymerization.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 441.54 |
| Purity | 98% |
| Molecular Weight (g/mol) | 441.542 |
| Chemical Name or Material | Troglitazone |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?