missing translation for 'onlineSavingsMsg'
Learn More

Tropathiane, Dr. Ehrenstorfer

Product Code. 30322855
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322855 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322855 Supplier Dr. Ehrenstorfer Supplier No. DREC17894920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tropathiane
CAS 67715-80-4
Analyte or Component Names Tropathiane
Shipping Condition +20°C
Molecular Formula C8 H16 O S
Certifications/Compliance ISO/IEC 17025
Synonym 2-Methyl-4-propyl-1,3-oxathiane, Tropathiane, 1,3-Oxathiane, 2-methyl-4-propyl-
InChI Formula InChI=1S/C8H16OS/c1-3-4-8-5-6-9-7(2)10-8/h7-8H,3-6H2,1-2H3
SMILES CCCC1CCOC(C)S1
IUPAC Name 2-methyl-4-propyl-1,3-oxathiane
Molecular Weight (g/mol) 160.277
Quantity 100 mg
Formula Weight 160.092 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.