missing translation for 'onlineSavingsMsg'
Learn More

Tropine-3-thiol Hydrochloride, TRC

Product Code. 30449189
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
30449189 25 g 25g
30451904 1 g -
30450419 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 30449189

Brand: Toronto Research Chemicals TRCT89267525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 908266-48-8
Formula Weight 193.0692
InChI Formula InChI=1S/C8H15NS.ClH/c1-9-6-2-3-7(9)5-8(10)4-6;/h6-8,10H,2-5H2,1H3;1H/t6-,7?,8+;/m0./s1
IUPAC Name (5S)-8-methyl-8-azabicyclo[3.2.1]octane-3-thiol;hydrochloride
Molecular Formula C8H16ClNS
Molecular Weight (g/mol) 193.74
SMILES Cl.CN1[C@H]2CCC1C[C@H](S)C2
Synonym (3-endo)-8-Methyl-8-Azabicyclo[3.2.1]octane-3-thiol Hydrochloride
Chemical Name or Material Tropine-3-thiol Hydrochloride

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.