missing translation for 'onlineSavingsMsg'
Learn More

Tropisetron Hydrochloride, TRC

Product Code. 30452327
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452327 10 mg -
30453142 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452327 Supplier Toronto Research Chemicals Supplier No. TRCT89270010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105826-92-4
Molecular Formula C17 H20 N2 O2 . Cl H
Molecular Weight (g/mol) 320.81
Synonym Tropisetron hydrochloride, (1R,3r,5S)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl 1H-indole-3-carboxylate hydrochloride, 1H-Indole-3-carboxylic acid, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (1:1), 1H-Indole-3-carboxylic acid, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, monohydrochloride (9CI), 1H-Indole-3-carboxylic acid, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, monohydrochloride, endo-, Novaban, SDZ-ICS 930, Tropisetron hydrochloride
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride
SMILES Cl.CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)c3c[nH]c4ccccc34

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H20N2O2.ClH/c1-19-11-6-7-12(19)9-13(8-11)21-17(20)15-10-18-16-5-3-2-4-14(15)16;/h2-5,10-13,18H,6-9H2,1H3;1H/t11-,12+,13+;
Formula Weight 320.1292
Chemical Name or Material Tropisetron hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.