missing translation for 'onlineSavingsMsg'
Learn More

Tropolone, TRC

Product Code. 30012514
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012514 50 g 50g
17910565 10 g 10g
17941925 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012514 Supplier Toronto Research Chemicals Supplier No. TRCT89270550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-75-5
Molecular Formula C7 H6 O2
Molecular Weight (g/mol) 122.121
Synonym 2-Hydroxy-2,4,6-cycloheptatrien-1-one, 2-Hydroxytropone, NSC 89303, Purpurocatechol, α-Tropolone
IUPAC Name 2-hydroxycyclohepta-2,4,6-trien-1-one
SMILES OC1=CC=CC=CC1=O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H6O2/c8-6-4-2-1-3-5-7(6)9/h1-5H,(H,8,9)
Formula Weight 122.0368
Percent Purity >95%
Chemical Name or Material Tropolone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.