Learn More
TRPC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129441
Description
PA1-29441 detects TRPC4 from rat and mouse samples. PA1-29441 is expected to react with human samples due to sequence homology. PA1-29441 has been successfully used in Western blot applications. Predicted molecular weight: 117 kDa. The PA1-29441 immunogen is a synthetic peptide: CKEKHAHEEDSSIDYDL, corresponding to amino acids 943-958 of mouse TRPC4.
TRPC4 is a member of the canonical subfamily of transient receptor potential cation channels. TRPC4 forms a non-selective calcium-permeable cation channel that is activated by Gq-coupled receptors and tyrosine kinases, and plays a role in multiple processes including endothelial permeability, vasodilation, neurotransmitter release and cell proliferation. Single nucleotide polymorphisms in TRPC4 may be associated with generalized epilepsy with photosensitivity. Alternatively spliced transcript variants encoding multiple isoforms have been observed for TRPC4. The classical or canonical transient receptor potential (TRPC) channels are the subfamily most closely related to the founding member of the TRP family, the Drosophila TRP channel. TRPC channels have seven TRPC genes that encode the channel protein (TRPC1-7). Diseases associated with TRPC4 include Photosensitive Epilepsy. CaV3.3 encodes the pore-forming alpha subunit of a voltage gated calcium channel. The encoded protein is a member of a subfamily of calcium channels referred to as is a low voltage-activated, T-type, calcium channel. The channel encoded by CaV3.3 is characterized by a slower activation and inactivation compared to other T-type calcium channels. CaV3.3 may be involved in calcium signaling in neurons. Alternate splicing results in multiple transcript variants of CaV3.3. Voltage-gated calcium channels (CaV) are present in the membrane of most excitable cells and mediate calcium influx in response to depolarization, an proteins such as CaV3.3 regulate intracellular processes such as contraction, secretion, neurotransmission and gene expression. Diseases associated with CACNA1I include Childhood Absence Epilepsy.
Specifications
| TRPC4 | |
| Polyclonal | |
| Unconjugated | |
| Q9UBN4, Q9QUQ5, O35119 | |
| TRPC4 | |
| synthetic peptide CKEKHAHEEDSSIDYDL, corresponding to amino acids 943-958 of Mouse TRPC4. | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.3 mg/mL | |
| TRPC4 | |
| Short transient receptor potential channel 4, Trp4, Capacitative calcium entry channel 1, CCE1. GENE NAME: Trpc4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22066, 7223, 84494 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.