missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 450.00€
Specifications
| Antigen | TRPM2 |
|---|---|
| Concentration | 1.5 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18270341
|
Novus Biologicals
NB110-82364SS |
0.025 mL |
213.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18780964
|
Novus Biologicals
NB110-82364 |
0.1 mL |
450.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRPM2 Polyclonal specifically detects TRPM2 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| TRPM2 | |
| Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cell Biology, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| 7226 | |
| Synthetic peptide made to an internal portion of the mouse TRPM2 protein (within residues 1200-1300). [Swiss-Prot# Q91YD4] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.5 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Mouse, Rat | |
| EC 3.6.1.13, EREG1MGC133383, Estrogen-responsive element-associated gene 1 protein, KNP3LTrpC-2, Long transient receptor potential channel 2, LTrpC2, LTRPC2TRPC7transient receptor potential cation channel subfamily M member 2, NUDT9H, NUDT9L1, transient receptor potential cation channel, subfamily M, member 2, Transient receptor potential channel 7, TrpC7 | |
| TRPM2 | |
| IgG | |
| Affinity Purified | |
| 172 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title