missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM4 Antibody (CL11214) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07990AF532
This item is not returnable.
View return policy
Beskrivning
TRPM4 Monoclonal antibody specifically detects TRPM4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifikationer
| TRPM4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Calcium-activated non-selective cation channel 1, FLJ20041, hTRPM4, Long transient receptor potential channel 4, LTrpC-4, LTRPC4, melastatin-4, PFHB1B, transient receptor potential cation channel subfamily M member 4, transient receptor potential cation channel, subfamily M, member 4, TRPM4B | |
| This antibody was developed using a recombinant protein derived from Q8TD43, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Endocrinology, Neuroscience, Signal Transduction | |
| 54795 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL11214 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering