Learn More
TRPV1 Polyclonal Antibody, OriGene
Rabbit Polyclonal Antibody
Brand:  Origene Technologies TA890016
Encompass_Preferred
 View more versions of this product
Product Code. 17599424
  • 492.47€ / 100µL
Description
TRPV1 (Transient Receptor Potential Cation Channel Subfamily V Member 1) is a receptor for capsaicin, the main pungent ingredient in hot chili peppers, and elicits a sensation of burning pain by selectively activating sensory neurons. TRPV1 is a non-selective cation channel that is structurally related to members of the TRP family of ion channels such as TRPC3 and TRPC6. TRPV1 is permeable to calcium, magnesium, and monovalent cations of sodium and potassium. Structurally, TRPV1 contains 6 trans-membrane domains, a P-loop membrane domain and 3 ankyrin repeat domains. TRPV1 is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo. TRPV1 seems to mediate proton influx and may be involved in intracellular acidosis in nociceptive neurons. The expression of TRPV1 appears to be limited to neurons within the sensory ganglia. Four transcript variants encoding TRPV1 with variations in th 5′ UTR sequence have been described. Diseases associated with TRPV1 include Irritable Bowel Syndrome (IBS) and Bladder Disease.
Specifications
Specifications
TRPV1
Polyclonal
PBS with 50% glycerol and 0.02% sodium azide
Q8NER1
TRPV1
Recombit protein of human TRPV1
100 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
TRPV1
VR1
Rabbit
Antigen Affinity Chromatography
RUO
7442
-20° C, Avoid Freeze/Thaw Cycles