missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPV5 Recombinant Rabbit Monoclonal Antibody (24GB1850)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557296
This item is not returnable.
View return policy
Description
TRPV5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene is a member of the transient receptor family and the TrpV subfamily. The calcium-selective channel encoded by this gene has 6 transmembrane-spanning domains, multiple potential phosphorylation sites, an N-linked glycosylation site, and 5 ANK repeats. This protein forms homotetramers or heterotetramers and is activated by a low internal calcium level.
Specifications
| TRPV5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TRPV5 | |
| Calciu; Calcium transport protein 2; calcium transporter 2; CaT2; D630033B11; ECAC; Ecac1; epithelial calcium channel; epithelial calcium channel 1; Osm-9-like TRP channel 3; OTRPC3; Transient receptor potential cation channel subfamily V member 5; transient receptor potential cation channel, subfamily V, member 5; Trpv5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116469, 194352, 56302 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1850 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P69744, Q9JIP0, Q9NQA5 | |
| TRPV5 | |
| A synthesized peptide derived from human TRPV5 (aa 1-100). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction