missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptamine Hydrochloride, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
Specifications
Specifications
| Chemical Name or Material | Tryptamine hydrochloride |
| CAS | 343-94-2 |
| Molecular Formula | C10 H12 N2 . H Cl |
| Quantity | 2.5 g |
| Synonym | 1H-Indole-3-ethanamine HCl |
| InChI Formula | InChI=1S/C10H12N2.ClH/c11-6-5-8-7-12-10-4-2-1-3-9(8)10;/h1-4,7,12H,5-6,11H2;1H |
| SMILES | Cl.NCCc1c[nH]c2ccccc12 |
| IUPAC Name | 2-(1H-indol-3-yl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 196.677 |
| Formula Weight | 196.0767 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction