missing translation for 'onlineSavingsMsg'
Learn More

Tryptamine Hydrochloride, TRC

Product Code. 30451089
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451089 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451089 Supplier Toronto Research Chemicals Supplier No. TRCT894603250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tryptamine hydrochloride
CAS 343-94-2
Molecular Formula C10 H12 N2 . H Cl
Quantity 250 mg
Synonym 1H-Indole-3-ethanamine HCl
InChI Formula InChI=1S/C10H12N2.ClH/c11-6-5-8-7-12-10-4-2-1-3-9(8)10;/h1-4,7,12H,5-6,11H2;1H
SMILES Cl.NCCc1c[nH]c2ccccc12
IUPAC Name 2-(1H-indol-3-yl)ethanamine;hydrochloride
Molecular Weight (g/mol) 196.677
Formula Weight 196.0767
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.