missing translation for 'onlineSavingsMsg'
Learn More

Tryptophol, TRC

Product Code. 30132995
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132995 5 g 5g
30012912 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132995 Supplier Toronto Research Chemicals Supplier No. TRCT8947005G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-55-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.2
Synonym Indole-3-ethanol, 2-(1H-Indol-3-yl)ethanol, 2-(3-Indolyl)ethanol, 3-(2-Hydroxyethyl)-1H-indole, 3-(2-Hydroxyethyl)indole, 3-(β-Hydroxyethyl)indole, IEA, NSC 3884, β-(3-Indole)ethanol;
IUPAC Name 2-(1H-indol-3-yl)ethanol
SMILES OCCC1=CNC2=C1C=CC=C2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H11NO/c12-6-5-8-7-11-10-4-2-1-3-9(8)10/h1-4,7,11-12H,5-6H2
Formula Weight 161.0841
Percent Purity >95%
Chemical Name or Material Tryptophol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.